Push-to-connect union elbow, 316 stainless steel, 1/4"
Representative image only.
Item # EW-31370-19
The ease of push-to-connect and the durability of 316 stainless steel
PRODUCT OPTIONS
$64.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Seal MaterialFKM
  • Sterilization MethodNot recommended
  • Connection TypePush-to-Connect
  • Push-to-Connect Tube OD (port 1) (in)1/4
  • Push-to-Connect Tube OD (port 2) (in)1/4
  • Min Temperature (° F)-4
  • Max Temperature (° F)300
  • Min Temperature (° C)-20
  • Max Temperature (° C)149
  • Max Operating Pressure (PSI)225
  • Max Vacuum (in Hg)30
  • Max Vacuum (mm Hg)762
  • DescriptionPush-to-connect union elbow, 316 stainless steel, 1/4"
More About this Item
The durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines. Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines. Matching material of construction to your stainless piping will reduce the effects of thermal
expansion differentials and minimize line leakage.