PVC Check Valve, 1/2" NPT(F) connections
Representative image only.
Item # EW-01350-24
Create a secure pipe threaded connection
  • Choose from PVDF, PVC, or polypropylene (PP) bodies
PRODUCT OPTIONS
$133.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Breaking Pressure (PSI)1.5
  • Cv8
  • Max Pressure (PSI)150
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, Viton
  • DescriptionPVC Check Valve, 1/2" NPT(F) connections
Key Features
  • Choose from PVDF, PVC, or polypropylene (PP) bodies
More About this Item
All valves have Viton® diaphragms.