PVC Stopcock w/ Flow Indication, 1/4-1/2" ID x 1/4-1/2" ID
Representative image only.
Item # EW-06225-56
Easily control flow with flow indicator handle
  • Constructed of durable PVC and EPDM
PRODUCT OPTIONS
$43.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" to 1/2" Barb
  • Connection TypeBarbed
  • Inlet Port1/4" to 1/2" Barb
  • Orifice (in)0.25 to 0.5
  • Orifice (mm)6.35 to 12.7
  • Max Pressure (PSI)150
  • Max Temperature (° F)120
  • Max Temperature (° C)48
  • Wetted MaterialsPVC
  • Tubing ID (in)0.25 to 0.5
  • DescriptionPVC Stopcock w/ Flow Indication, 1/4-1/2" ID x 1/4-1/2" ID
Key Features
  • Constructed of durable PVC and EPDM
More About this Item
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120°>F (49°>C); maximum vacuum is 29.9'' Hg.