Pyrex 3025-50 Cylinder, Brand 3025, graduated, 50 mL, case of 18
Representative image only.
Pyrex Mfr # 3025-50 – Item # EW-34546-23
Double metric scale, economy
PRODUCT OPTIONS
$450.00USD / Case of 18
BUY MORE & SAVE
  • 1-4$450.00USD / Case of 18
  • 5-9$405.00USD / Case of 18
  • 10+$382.50USD / Case of 18
$89.50 - $162.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)50
  • Graduation Interval (mL)1
  • DescriptionCylinder, Brand 3025, graduated, 50 mL, case of 18
More About this Item
These double metric scale cylinders are calibrated "to contain." These economical cylinders are ideal for many general laboratory procedures.
Graduated ware must carry markings listing its capacity and the temperature at which it should be used, and whether the piece was calibrated T.C., "to contain," or T.D., "to deliver," the stated volume. T.C. calibration indicates the volume held in the co