Pyrex 3025-500 Cylinder, Brand 3025, graduated, 500 mL, case of 8
Representative image only.
Pyrex Mfr # 3025-500 – Item # EW-34546-26
Double metric scale, economy
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)500
  • Graduation Interval (mL)5
  • DescriptionCylinder, Brand 3025, graduated, 500 mL, case of 8
More About this Item
These double metric scale cylinders are calibrated "to contain." These economical cylinders are ideal for many general laboratory procedures.
Graduated ware must carry markings listing its capacity and the temperature at which it should be used, and whether the piece was calibrated T.C., "to contain," or T.D., "to deliver," the stated volume. T.C. calibration indicates the volume held in the co