Pyrex Brand 5642 Class A Volumetric Flask, 100 mL; 6/PK
Representative image only.
Pyrex Mfr # 5642-100 – Item # EW-34561-06
No more broken stoppers
  • High-density polyethylene stopper won’t break if dropped
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 100 mL capacity with ±0.08 mL tolerance
  • Includes no.13 glass stopper
PRODUCT OPTIONS
$410.00USD / Pkg of 6
BUY MORE & SAVE
  • 1-4$410.00USD / Pkg of 6
  • 5-9$369.00USD / Pkg of 6
  • 10+$348.50USD / Pkg of 6
$145.00 - $218.00USD / Each
for an additional
$145.00USD / Each
for an additional
$218.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)100
  • Tolerance (mL)±0.08
  • Cap TypePlastic stopper
  • Cap MaterialHDPE
  • Stopper Number Accepted13
  • Overall Height (cm)18.1
  • DescriptionBrand 5642 Volumetric Flask; 100 mL, pack of 6
Key Features
  • High-density polyethylene stopper won’t break if dropped
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 100 mL capacity with ±0.08 mL tolerance
  • Includes no.13 glass stopper
More About this Item
These PYREX® Brand 5642 Class A volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks to provide excellent durability. Volumetric flasks include a linear high-density polyethylene stopper that conforms to stopper dimensions. The graduation line is sharp and permanent. Large white block letters make the inscriptions easy to read. The 2-mL flask is test tube shaped.
When taper sizes are given, the first number is the stopper diameter (mm) across the top; the second number is the stopper length (mm). If only one number is given, it indicates the stopper diameter (mm) across the top.