Pyrex 5642-2L Brand 5642 Volumetric Flask; 2000 mL, pack of 1
Representative image only.
Pyrex Mfr # 5642-2L – Item # EW-34561-11
No more broken stoppers
  • High-density polyethylene stopper won’t break if dropped
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 2000 mL capacity with ±0.50 mL tolerance
  • Includes no.27 glass stopper
PRODUCT OPTIONS
$139.00USD / Pkg of 1
BUY MORE & SAVE
  • 1-4$139.00USD / Pkg of 1
  • 5-9$125.10USD / Pkg of 1
  • 10+$118.15USD / Pkg of 1
$70.00 - $139.00USD / Each
for an additional
$70.00USD / Each
for an additional
$139.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)2000
  • Tolerance (mL)±0.5
  • Cap TypePlastic stopper
  • Cap MaterialHDPE
  • Stopper Number Accepted27
  • Overall Height (cm)38.8
  • DescriptionBrand 5642 Volumetric Flask; 2000 mL, pack of 1
Includes
volumetric flask and stopper.
Key Features
  • High-density polyethylene stopper won’t break if dropped
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 2000 mL capacity with ±0.50 mL tolerance
  • Includes no.27 glass stopper
More About this Item
These PYREX® Brand 5642 Class A volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks to provide excellent durability. Volumetric flasks include a linear high-density polyethylene stopper that conforms to stopper dimensions. The graduation line is sharp and permanent. Large white block letters make the inscriptions easy to read. The 2-mL flask is test tube shaped.
When taper sizes are given, the first number is the stopper diameter (mm) across the top; the second number is the stopper length (mm). If only one number is given, it indicates the stopper diameter (mm) across the top.