Pyrex Brand 5660 Lifetime Red™ Class A Volumetric Flask; 100 mL; 6/PK
Representative image only.
Pyrex Mfr # 5660-100 – Item # EW-34562-03
No more worn off graduations
  • Get permanent legibility and accuracy with etched red graduation
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 100 mL capacity with ±0.08 mL tolerance
  • Includes no.13 glass stopper
PRODUCT OPTIONS
$405.00USD / Pkg of 6
BUY MORE & SAVE
  • 1-4$405.00USD / Pkg of 6
  • 5-9$364.50USD / Pkg of 6
  • 10+$344.25USD / Pkg of 6
$145.00 - $218.00USD / Each
for an additional
$145.00USD / Each
for an additional
$218.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)100
  • Tolerance (mL)±0.08
  • Cap TypeGlass stopper
  • Cap MaterialGlass
  • Stopper Number Accepted13
  • Overall Height (cm)18.2
  • DescriptionBrand 5660 Lifetime Red Volumetric Flask; 100 mL, pack of 6
Key Features
  • Get permanent legibility and accuracy with etched red graduation
  • Volumetric flasks are constructed of borosilicate glass
  • Volumetric flask features 100 mL capacity with ±0.08 mL tolerance
  • Includes no.13 glass stopper
More About this Item
These PYREX® Brand 5660 Class A volumetric flasks feature a fine line capacity mark etched through a narrow red band into clear glass, making a filler unnecessary and providing high legibility, accuracy, and permanence.
The strength of these flasks has been increased appreciably through machine-blown bodies that feature sealed heavy-tubing necks.
Flasks are calibrated "To Contain" and meet Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.