Qorpak 2303 Boston Bottle Mailer, 8 oz, 6-1/2" L x 2-1/2" dia, 24/cs

Representative image only.
Qorpak Mfr # GLC-01172 – Item # EW-06148-42
Meet government regulations for mailing liquids

$302.00USD / Case of 24

BUY MORE & SAVE

  • 1-5$302.00USD / Case of 24
  • 6-29$271.80USD / Case of 24
  • 30-119$256.70USD / Case of 24
  • 120+$241.60USD / Case of 24

SPECS & DESCRIPTION

Specifications & Description

  • MaterialFiberboard
  • Fits Bottle / Beaker Size (mL)250
  • Cap MaterialMetal
  • AutoclavableNo
  • Outside Diameter (OD) (in)2 1/2
  • Outside Diameter (OD) (cm)6.4
  • Overall Height (in)6 1/2
  • Overall Height (cm)16.5
  • DescriptionBoston Bottle Mailer, 8 oz, 6-1/2" L x 2-1/2" dia, 24/cs

More About this Item

Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat