QuickSip 4724150 Optional bottletop aspirator
Representative image only.
QuickSip Mfr # 4724150 – Item # EW-07912-40
Remove supernatants without an external vacuum or power supply
  • Portable and easy to use at multiple workstations
  • Mounts on most reagent bottles with 45 mm thread with 45/33 mm and 45/38 mm adapters (included)
  • Compatible with disposable pipette tips, micropipettes, and Pasteur pipettes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Wetted PartsPolypropylene, polyethylene and silicone
  • DescriptionOptional bottletop aspirator
  • Warranty1 year
1 Year Warranty
Key Features
  • Portable and easy to use at multiple workstations
  • Mounts on most reagent bottles with 45 mm thread with 45/33 mm and 45/38 mm adapters (included)
  • Compatible with disposable pipette tips, micropipettes, and Pasteur pipettes
  • 1 year warranty
More About this Item
This bottletop aspirator conveniently removes supernatants from dishes, flasks, culture bottles, multiwell plates, and more. Ideal for removing liquid from centrifuge tubes after precipitation of proteins and nucleic acids. Use the optional eight-channel aspirator manifold for quick aspiration of multiwell plates.
Aspiration is fingertip controlled with each plunger stroke powering up to 25 mL of supernatant. Replaceable piston/cylinder cartridge allows you to quickly switch to a new supernatant without mixing aspirated fluids.
What’s included: QuickSip™ controller, 73" (200 cm) of silicone tubing, 1 spare piston/cylinder cartridge, polypropylene thread adapters (GL 45⁄33 and GL 45⁄38) and instruction manual.