QV2-186-M Diaphragm Valve, PFA, 1/2" NPT(F), 75 psi max
Representative image only.
Mfr # QV2-186-M – Item # EW-30502-07
$504.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Orifice (in)0.375
  • Orifice (mm)9.53
  • Cv1.17
  • DescriptionDiaphragm Valve, PFA, 1/2" NPT(F), 75 psi max
More About this Item
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.