RAE Systems 010-0901-000 Manual gas detection pump, 50-100 cc sample size
Representative image only.
RAE Systems Mfr # 010-0901-000 – Item # EW-86260-00
Springless design for accurate 50- and 100-cc sample volumes
  • Flow finish indicator signals stroke completion
  • Built-in tube tip breaker for clean break every time
$232.90USD / Each
$274.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionManual gas detection pump, 50-100 cc sample size
Key Features
  • Flow finish indicator signals stroke completion
  • Built-in tube tip breaker for clean break every time
More About this Item
Hand-operated precision piston pump works with RAE System gas detection tubes (sold separately). Features flow-finish indicator to signal stroke completion, stroke counter, and built-in tube-tip breaker. Includes wrist strap.