RAE Systems 10-103-10-4LL Gas Detection Tubes, Hydrogen Sulfide (H2S), 2.5 to 60 ppmv; 10/Box
Representative image only.
RAE Systems Mfr # H10-103-10 – Item # EW-86260-62
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredHydrogen sulfide
  • Range2.5 to 60 ppm
  • TLV (ppm)10
  • DescriptionGas Detection Tubes, Hydrogen Sulfide (H2S), 2.5 to 60 ppmv; 10/Box
  • Warranty1 year
1 Year Warranty
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 1 year warranty
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.