RAE Systems 10-103-30-4H Gas detection tubes, hydrogen sulfide, 100 to 2000ppm, 10/box
Representative image only.
RAE Systems Mfr # H10-103-30 – Item # EW-86260-79
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
PRODUCT OPTIONS
$46.75USD / Pkg of 10
$55.00USD / Pkg of 10
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredHydrogen sulfide
  • Range100 to 2000
  • DescriptionGas detection tubes, hydrogen sulfide, 100 to 2000ppm, 10/box
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.