RAE Systems 10-103-40-4HH Gas detection tubes, hydrogen sulfide, 0.1 to 2%, 10/box

Representative image only.
RAE Systems Mfr # 10-103-40-4HH – Item # EW-86260-64
Sharp, clear color change for unambiguous measurements

PRODUCT OPTIONS

$39.31USD / Pkg of 10
$46.25USD / Pkg of 10
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Gas MeasuredHydrogen sulfide
  • Range0.1 to 2%
  • TLV (ppm)10
  • DescriptionGas detection tubes, hydrogen sulfide, 0.1 to 2%, 10/box

More About this Item

Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat