RAE Systems 10-104-30-2LL Gas detection tubes, carbon dioxide, 300 to 5000 ppm, 10/box
Representative image only.
RAE Systems Mfr # H10-104-30 – Item # EW-86260-70
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredCarbon dioxide
  • Range300 to 5000
  • DescriptionGas detection tubes, carbon dioxide, 300 to 5000 ppm, 10/box
  • Warranty2 year
Two year warranty
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.