RAE Systems 10-108-10-14L Gas detection tubes, hydrogen chloride, 1 to 20 ppm, 10/box
Representative image only.
RAE Systems Mfr # H10-108-10 – Item # EW-86260-74
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredHydrogen chloride
  • Range1 to 10
  • DescriptionGas detection tubes, hydrogen chloride, 1 to 20 ppm, 10/box
  • Warranty2 year
Two year warranty
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.