RAE Systems 10-113-20-152 Gas detection tubes, MEK, 0.02 to 6%, 10/box
Representative image only.
RAE Systems Mfr # H10-113-20 – Item # EW-86260-80
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
PRODUCT OPTIONS
$44.20USD / Pkg of 10
$52.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredMethyl Ethyl Ketone (MEK)
  • Range0.02 to 6%
  • DescriptionGas detection tubes, MEK, 0.02 to 6%, 10/box
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.