RAE Systems 10-116-10-7 Gas detection tubes, phosphine, 5 to 50 ppm, 10/box
Representative image only.
RAE Systems Mfr # H10-116-10 – Item # EW-86260-81
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
PRODUCT OPTIONS
$42.28USD / Pkg of 10
$49.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredPhosphine
  • Range5 to 50
  • DescriptionGas detection tubes, phosphine, 5 to 50 ppm, 10/box
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.