Ralston Instruments QSCM-5KPSIG-D Calibration Manifold w/Digital Guage; 1/4" NPTF/SS
Representative image only.
Ralston Instruments Mfr # QSCM-5KPSIG-D – Item # EW-68690-14
Quickly and economically calibrate high-pressure instrumentation using nitrogen
  • Proprietary pressure balanced fine adjustment piston allows for precise control of pressure up to 3000 psi (207 bar)
  • Fine adjust piston, fill, balance and vent valves are in one light weight, compact unit
  • Soft-seated valves make filling and venting the system very precise
  • Low volume Quick-test hose, gauge adapter and process connection can be attached without thread sealant or a wrench
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCalibration Manifold w/Digital Guage; 1/4\" NPTF/SS
  • Warranty1 Year
1 Year Warranty
Key Features
  • Proprietary pressure balanced fine adjustment piston allows for precise control of pressure up to 3000 psi (207 bar)
  • Fine adjust piston, fill, balance and vent valves are in one light weight, compact unit
  • Soft-seated valves make filling and venting the system very precise
  • Low volume Quick-test hose, gauge adapter and process connection can be attached without thread sealant or a wrench
  • 1 year warranty
More About this Item
The most cost effective way to perform pressure calibration with one compact, lightweight unit. Easy to use — just quickly attach or detach reference with one pressure gauge, calibrator or pressure module connection. A fine adjustment piston allows precise pressure control. The Delrin® acetal, soft-seated fill and bleed valves allow for small adjustments to pressure. Also included are low volume hoses which conserve nitrogen in the cylinder so it lasts longer. The stainless steel calibration manifolds are ideal for remote field calibration to 5000 psi (345 bar).
Includes nylon bag, 1/4" Female NPT - gauge adapter, 1/4" Male NPT process adapter, digital 5000psi,±0.25%FS gauge, 3ft. & 6ft. quick-test hoses.