Rapid Bacterial Test Kit, Salmonella, anti-body based, 20 tests/kit
Representative image only.
Item # EW-36151-00
Fast and reliable tests for detecting and identifying bacteria
  • Not available for sale outside of the United States due to shipping constraints
  • Not for sale outside the U.S. due to shelf life and shipping restrictions
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeSalmonella
  • Number of Tests Included20
  • DescriptionRapid Bacterial Test Kit, Salmonella, anti-body based, 20 tests/kit
Key Features
  • Not available for sale outside of the United States due to shipping constraints
  • Not for sale outside the U.S. due to shelf life and shipping restrictions
More About this Item
These test kits provide a quick and easy way to determine the presence of different types of bacteria in food samples, environmental samples and sample colonies. All tests kits are simple to use and produce easy to identify positive or negative results. Kits contain everything to test cultured bacterial colonies. For lab use only.