Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Representative image only.
Item # EW-06833-00
Won't rust, corrode, or distort
  • Constructed of durable Celcon plastic
PRODUCT OPTIONS
$128.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Pinch Valve Tube Size1/8" to 1/2" OD
  • Housing MaterialPolyoxymethylene (POM) Acetal
  • DescriptionRatchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Key Features
  • Constructed of durable Celcon plastic
More About this Item
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.