Reagents Formic Acid, 90%, 4L, Viscosity Grade

Representative image only.
Reagents Mfr # C2105050-4A – Item # EW-78923-91
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Low Iron
  • 3 year warranty

$197.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeViscosity
  • Linear FormulaHCOOH
  • ClassAcid
  • ChemistryAcids
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight46.03
  • Vapor Pressure (torr)30.0025
  • PackagingPoly Bottle
  • DOT InformationFORMIC ACID, 8 (3)
  • UN NumberUN1779
  • CAS Number64-18-6
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionFormic Acid, 90%, 4L, Viscosity Grade
  • Warranty3 year
3 Year Warranty

Key Features

  • Low Iron
  • 3 year warranty

More About this Item

Formic Acid, 4L, 90% ConcentratIon, Viscosity Grade, Colorless, Liquid, 101°C Boiling Point, 50°C Flash Point, <2 pH Range, 1.22 Density, 36 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat