Reagents Hydrochloric Acid, 0.20N, 1L, Standardized
Representative image only.
Reagents Mfr # C7101020-1A – Item # EW-78915-66
Manufactured using the highest quality raw materials in ISO 9001 facility
  • N/50, Assay 0.1995 to 0.2005 Normal, Aqueous
  • 2 year warranty
$34.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Normality0.2
  • ClassStandardized
  • ChemistryAcids
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionHydrochloric Acid, 0.20N, 1L, Standardized
  • Warranty2 year
Two year warranty
Key Features
  • N/50, Assay 0.1995 to 0.2005 Normal, Aqueous
  • 2 year warranty
More About this Item
Hydrochloric Acid, 1L, 0.2N ConcentratIon, Colorless, Liquid, 100°C Boiling Point, 0°C Melting Point, <1 pH Range, 1 Density, 24 Months Shelf Life.