Reagents Kallings Reagent I, 500mL

Representative image only.
Reagents Mfr # CK157000-500A – Item # UX-78911-77
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Reagents and solutions to meet your lab and research needs
  • 2 year warranty

$210.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassMixture
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationCORROSIVE LIQUIDS, FLAMMABLE, N.O.S. (HYDROCHLORIC ACID, ETHANOL), 8 (3)
  • UN NumberUN2920
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionKallings Reagent I, 500mL,
  • Warranty2 year
Two year warranty

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat