Reagents Nitric Acid 70% Trace Metal, SC, 2.5L , ACS

Representative image only.
Reagents Mfr # C5101650-2.5D1 – Item # EW-78930-14
Manufactured using the highest quality raw materials in ISO 9001 facility
  • 68.00-70.00%
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeTrace Metals ppm
  • Linear FormulaHNO3
  • ClassAcid
  • ChemistryAcids
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight63.0128
  • Vapor Pressure (torr)7.0506
  • PackagingClear Glass Bottle
  • DOT InformationNITRIC ACID, 8 (5.1)
  • CAS Number7697-37-2
  • UN NumberUN2031
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionNitric Acid 70% Trace Metal, SC, 2.5L, ACS
  • Warranty3 year
3 Year Warranty

Key Features

  • 68.00-70.00%
  • 3 year warranty

More About this Item

Nitric Acid, 2.5L, 70% Concentration, Trace Metal Grade, Colorless, Liquid, 121°C Boiling Point, -32°C Melting Point, <1 pH Range, 1.416 Density, 37 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat