Reagents Sodium Carbonate Anhydrous, 2.5kg, Purified

Representative image only.
Reagents Mfr # C2312100-2.5A5 – Item # EW-78919-35
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Granular
  • 10 year warranty

$60.70USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePurified
  • Linear FormulaNa2CO3
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight105.99
  • PackagingPoly Jar
  • CAS Number497-19-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Carbonate Anhydrous, 2.5kg, Purified
  • Warranty10 year
10 Year Warranty

Key Features

  • Granular
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat