Reagents Sodium Chloride, Purified, Wide-Mouth Container, 500 g

Representative image only.
Reagents Mfr # C2313000-500A5 – Item # EW-78913-69
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white crystals
  • Density: 2.17
  • Solubility information: 360 g/L at 25 °C (68 °F)
  • pH range: 5 to 9
  • 500 g wide-mouth container
  • 20 years warranty

PRODUCT OPTIONS

$20.20USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePurified
  • Linear FormulaNaCl
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight58.44
  • Vapor Pressure (torr)1
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7647-14-5
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)1461
  • Boiling Point F (° F)2661.8
  • Melting Point C (° C)801
  • Melting Point F (° F)1473.8
  • DescriptionSodium Chloride, Purified, Wide-Mouth Container, 500 g
  • Warranty20 years
20 Year Warranty

Key Features

  • Colorless/white crystals
  • Density: 2.17
  • Solubility information: 360 g/L at 25 °C (68 °F)
  • pH range: 5 to 9
  • 500 g wide-mouth container
  • 20 years warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat