Reagents Sodium Persulfate, Reagent, Wide-Mouth Container, 500 g

Representative image only.
Reagents Mfr # C2323100-500A5 – Item # EW-78914-01
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white solid
  • Density: 2.6
  • Solubility information: 556 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container
  • 5 year warranty

PRODUCT OPTIONS

$41.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeReagent
  • Linear FormulaNa2S2O8
  • ClassSalts
  • FormSolid
  • SynonymsSodium peroxydisulfate
  • Weight (g)500
  • Formula Weight238.1
  • PackagingWide-mouth container
  • DOT InformationSODIUM PERSULFATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number7775-27-1
  • UN NumberUN1505
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)120
  • Melting Point F (° F)248
  • DescriptionSodium Persulfate, Reagent, Wide-Mouth Container, 500 g
  • Warranty5 years
5 Year Warranty

Key Features

  • Colorless/white solid
  • Density: 2.6
  • Solubility information: 556 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat