Reagents Sodium , Standard, 500mL, Standard
Representative image only.
Reagents Mfr # CS022600-500A – Item # EW-78913-50
Manufactured using the highest quality raw materials in ISO 9001 facility
  • 100ppm Na, Range 99.5 to 100.5ppm
  • 1 year warranty
$48.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ClassStandard
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • Shelf Life (days)380
  • Shelf Life (months)12
  • DescriptionSodium, Standard, 500mL, Standard
  • Warranty1 year
1 Year Warranty
Key Features
  • 100ppm Na, Range 99.5 to 100.5ppm
  • 1 year warranty
More About this Item
Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.