Reagents Sulfamic Acid, 500g, ACS Reagent

Representative image only.
Reagents Mfr # C2114300-500A5 – Item # EW-78914-29
Manufactured using the highest quality raw materials in ISO 9001 facility

$51.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS
  • Linear FormulaNH2SO3H
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)500
  • Formula Weight97.09
  • Vapor Pressure (torr)0.00585
  • PackagingPoly Jar
  • DOT InformationSULFAMIC ACID, 8
  • UN NumberUN2967
  • CAS Number5329-14-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSulfamic Acid, 500g, ACS Reagent

More About this Item

Sulfamic Acid, 100g, Practical Grade, 5329-14-6 CAS Number, NH2SO3H Chemical Formula, 97.09g/mol Molecular Weight, White to Off-white, Solid, 205°C Melting Point, 2.15 Density, 60 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat