Reagents Water, Reagent Grade, 4 L

Representative image only.
Reagents Mfr # C2346300-4A – Item # EW-78922-51
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Water meets ASTM D1193-06 Type I certification
  • 1 year warranty

$53.10USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeReagent
  • Empirical FormulaH2O
  • FormLiquid
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • Volume (mL)4000
  • Formula Weight18.02
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, Reagent Grade, 4 L
  • Warranty1 year
1 Year Warranty

Key Features

  • Water meets ASTM D1193-06 Type I certification
  • 1 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat