Red Lion 509FX N-Tron Unmanaged Industrial Ethernet Switch, 9 Port; SC
Representative image only.
Red Lion Mfr # 509FX-SC – Item # EW-17702-22
Ideal for connecting Ethernet-enabled industrial or security equipment
  • Unmanaged switches with full IEEE 802.3 and 1613 compliance
  • RJ-45 ports support full/half duplex operation and auto-sense duplex, speed, and MDIX
  • Redundant power inputs (10 to 30 VDC) with ESD and surge protection diodes on all ports
  • Compact footprint conserves space and is DIN-rail mountable
  • Bi-color LED lights indicate link, speed, activity, and duplex status
  • High MTBF — greater than 2M hours — ideal for harsh conditions
  • Store-and-forward and plug-and-play technology for ease of use
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Ports9
  • Max Input Voltage (VDC)30
  • Min Input Voltage (VDC)10
  • Max Input Current (mA)260
  • DescriptionN-Tron Unmanaged Industrial Ethernet Switch, 9 Port; SC
  • Warranty3 Years
3 Year Warranty
Key Features
  • Unmanaged switches with full IEEE 802.3 and 1613 compliance
  • RJ-45 ports support full/half duplex operation and auto-sense duplex, speed, and MDIX
  • Redundant power inputs (10 to 30 VDC) with ESD and surge protection diodes on all ports
  • Compact footprint conserves space and is DIN-rail mountable
  • Bi-color LED lights indicate link, speed, activity, and duplex status
  • High MTBF — greater than 2M hours — ideal for harsh conditions
  • Store-and-forward and plug-and-play technology for ease of use