Red Lion 68485-73 + 68485-44 5 Digit Display with MPAXP010 Input Module
Representative image only.
Red Lion Mfr # 68485-73 + 68485-44 – Item # EW-68495-10
Improve process visability and shop floor productivity
  • LPAX modules feature large 1.5" (38 mm) red display readable from 70 feet (21 meters)
  • EPAC modules feature extra-large 4" (101 mm) red display readable from 180 feet (55 meters)
  • User programmable inputs
  • Field-installable option cards
  • NEMA 4X / IP65-rated front panels
PRODUCT OPTIONS
$738.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description5 Digit Display with MPAXP010 Input Module
Key Features
  • LPAX modules feature large 1.5" (38 mm) red display readable from 70 feet (21 meters)
  • EPAC modules feature extra-large 4" (101 mm) red display readable from 180 feet (55 meters)
  • User programmable inputs
  • Field-installable option cards
  • NEMA 4X / IP65-rated front panels
More About this Item
Combining an MPAX input module with a large (LPAX) or extra-large (EPAX) display gives you a display size meter with the same functionality as the analog input PAX meters described on page 787. These large displays make an excellent productivity tool by providing your shop floor with easy to read plant status in real time.
We list below some of the more popular models with analog input, but others are available so check with our applications representative if you have a specific need not addressed. These diplays are field upgradeable with option cards shown below, including communications, setpoints and retransmitted analog outputs to provide flexibility and affordability as your needs grow.