This item is now discontinued. Please browse similar items
Red Lion 9000-PM Universal DIN/Panel/Rack Mount Kit for Switches
Representative image only.
Red Lion Mfr # 9000-PM – Item # EW-68586-01
Provides a wireless connection where installation of cabling is prohibitive
  • One 10/100BaseTX RJ-45 copper port
  • Three antennas for 3x3 MIMO operations
  • Four-user definable LED’s for display of signal quality
  • Radio enable, link/activity, and power LEDs
  • 802.3af PoE powered device
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUniversal DIN/Panel/Rack Mount Kit for Switches
Key Features
  • One 10/100BaseTX RJ-45 copper port
  • Three antennas for 3x3 MIMO operations
  • Four-user definable LED’s for display of signal quality
  • Radio enable, link/activity, and power LEDs
  • 802.3af PoE powered device
  • Autosensing 10/100BaseTX, duplex, and MDIX
  • Web browser management
  • Mutliple wireless modes—station, station WDS, access point, access point WDS
  • Mutliple network modes—bridge, router
  • 802.11i with AES-CCM & TKIP encryption and 802.1x, 64/128 bit WEP security
  • Legacy 802.11a/b/g (1-54Mbps) and 8802.11n (up to 300Mbps) data rates
  • Antenna-dependent range performance--indoor >300 m, outdoor >60 km
  • Redundant power inputs (20 to 49 VDC)
  • Rugged DIN-rail enclosure
  • Full IEEE 802.11 a, b, g, n compliance
More About this Item
With the proliferation of Ethernet in the industrial environment, situations arise where the installation of fiber or Cat5e cables and associated power cables is inconvenient, difficult, or cost prohibitive. The N-TRON™ 702-W industrial Ethernet device provides a wireless connection that can be quickly and easily deployed for these applications. It is ideally suited for connecting wireless devices to a wired network or for connecting two wired networks where it is not possible, impractical, or too expensive to install cable.
Three antennas enable the 702-W to utilize Multiple In, Multiple Out (MIMO) technology for increased throughput. Power over Ethernet (PoE) technology allows the 702-W to receive power through the Cat5e cable from a PoE-sourcing device, such as the N-TRON 105TX-POE switch or the 100-PoE4 midspan injector. Temporary deployments of network nodes are much easier using wireless and PoE technology because only one Cat5e cable is required.