Red Lion CSDIO14R Modular Controller 8 Inputs 6 Relay Out
Representative image only.
Red Lion Mfr # CSDIO14R – Item # EW-68483-93
SPECS & DESCRIPTION
Specifications & Description
  • Control Output 1Relay (x6)
  • Communication OutputEthernet
  • DescriptionModular Controller 8 Inputs 6 Relay Out
More About this Item
Red Lion’s Modular Controller series offers a cost effective solution to integrating multi-zone PID control, data acquisition and I/O into your PC, DCS or PLC control system. Users drag and drop controller data to PLC registers in seconds using point-and-click data mapping from an extensive menu of built-in serial and Ethernet drivers. There is no tedious PLC code to write. Instantly Ethernet-enable your PLC for free! Not only are programming costs slashed, you can eliminate costly add-on I/O and analog cards and solve applications simply using an inexpensive brick PLC. The high-density DIN rail design saves panel space and eliminates cutouts. All modules can be hot-swapped - the master automatically programs a replaced module.