Red Lion CUB5TB00 Timer, Red/Green Display Backlight Display
Representative image only.
Red Lion Mfr # CUB5TB00 – Item # EW-68510-14
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Display TypeLCD
  • Max Power VDC (VDC)28
  • Min Power VDC (VDC)10
  • EnclosureNEMA 4X; IP65
  • DescriptionTimer, Red/Green Display Backlight Display
  • Warranty2 Years
Two year warrantyConformite Europeenne
Key Features
  • 2 year warranty
More About this Item
The CUB5T features two input signals and eight operating modes to solve various Timing applications. Bi-directional timing with up to 18 different timing modes expand the unit’s capability.
  • 7 Digit Timing Capability
  • 6 Digit Cycle Counting Capability
  • Automatic Timer Reset Modes
    Extra Power
    The CUB5 Series can be powered by AC using an optional Red Lion Micro-Line/Sensor Power Supply that attaches directly to the back of any model. The MLPS1 is powered from an 85 to 250 VAC source and provides up to 400 mA to drive the CUB5 unit and any attached sensors.
    Communications Module
    Available in RS232 and RS485 formats and provides serial ASCII communications for sending and receiving data through the meter.
    Single Relay Output Module
    Features a Form C relay capable of switching up to 1 amp. Also programs to change backlight display when a setpoint is reached.
    Dual Transistor Module
    This module has two NPN-OC Transistors providing upper and lower setpoint control. Each output is capable of sinking 100 mA. This module operates with any of the CUB5 Analog Models.
  • REVIEWS