Red Lion G310S210 HMI 10.4" TFT 32K VGA 640x480 LCD, Outdoor
Representative image only.
Red Lion Mfr # G310S210 – Item # EW-30018-52
Benchmark of reliability and durability
  • Models come standard with one 10/100Base-T(X) Ethernet, two RS-232, and one RS-422/485 serial ports.
  • Communicates in over 300 different protocols.
  • Built-in data loggers for continuous data, event, and alarm collection and monitoring.
  • Data is compiled in CSV-formated files and can be stored, displayed, emailed, or automatically synchronized with a server.
  • A web server is built-in to allow remote control and monitoring.
$3,832.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHMI 10.4" TFT 32K VGA 640x480 LCD, Outdoor
Key Features
  • Models come standard with one 10/100Base-T(X) Ethernet, two RS-232, and one RS-422/485 serial ports.
  • Communicates in over 300 different protocols.
  • Built-in data loggers for continuous data, event, and alarm collection and monitoring.
  • Data is compiled in CSV-formated files and can be stored, displayed, emailed, or automatically synchronized with a server.
  • A web server is built-in to allow remote control and monitoring.
  • Programming via Red Lion Crimson 3.0 software platform.
  • 32-bit ColdFire processor
  • Compact flash socket for data logging
  • NEMA 4X / IP66 front panel
  • 24 VDC operation
  • 32 MB non-volitile flash memory
  • Dedicated menu key and 7 soft-keys
  • Resistive analog touchscreen