This item is now discontinued. Please browse similar items
Red Lion MPEC000 Magnetic Pickup Logic Emitter Follower 3-Pin Connector
Representative image only.
Red Lion Mfr # LMPEC000 – Item # EW-68485-83
SPECS & DESCRIPTION
Specifications & Description
  • Max Supply Voltage (VDC)30
  • Min Supply Voltage (VDC)9
  • Operating Frequency10 KHz max
  • Max Operating Temperature (° F)140
  • Min Operating Temperature (° F)0
  • Max Operating Temperature (° C)60
  • Min Operating Temperature (° C)-18
  • DescriptionMagnetic Pickup Logic Emitter Follower 3-Pin Connector
More About this Item
Logic Magnetic Pickups (LMPs) provide sensing sensitivities several orders of magnitude greater than standard magnetic pickups when detecting moving ferrous targets. By locating a high gain pre-amplifier & signal processing circuit "inside"the mag pickup housing, larger airgaps and slower target surface speeds can be achieved thereby greatly expanding application versatility as compared with conventional mag pickups. In addition, these units have excellent noise immunity and a frequency response to 10 KHz. The stainless steel sensing pole is mounted flush to the plastic sensing face allowing greater ease of setting airgaps and eliminating snagging the pole with a moving target.
Two output types are available (see ordering information). The NPN Open Collector Transistor Output unit provides a negative going current sinking output with the approach of a ferrous target and is current limited to 40 mA. The Transistor Emitter-Follower Output unit provides positive going 5 V pulses with the approach of a ferrous target and can source 20 mA of load current. The Open Collector units are available with either an integrally potted 10, 25 or 50 foot 3-wire shielded cable with neoprene strain relief boot or a polarized 3-pin 5/8"-24 threaded connector for quick change versatility. The Emitter-Follower output unit is available in the 3-pin connector version only.
FEATURES
  • Logic magnetic pickup
  • detects steel sensing gears or other ferrous targets
  • Built-in amplifier for low-speed operation
  • Stainless steel case
  • Epoxy encapsulated for oil, dirt and moisture resistance
  • 9 to 30 VDC powered
  • REVIEWS