Red Lion PAXLVD00 DC Voltage Panel Meter, 85-250 VAC
Representative image only.
Red Lion Mfr # PAXLVD00 – Item # EW-68486-17
SPECS & DESCRIPTION
Specifications & Description
  • DIN Size1/8\"
  • Display TypeLED
  • Max Power VAC (VAC)240
  • Min Power VAC (VAC)100
  • EnclosureNEMA 4X; IP65
  • DescriptionDC Voltage Panel Meter, 85-250 VAC
More About this Item
FEATURES
  • Seven-segment LED display
  • 1/8 DIN (50mm x 97mm)
  • Built-in scaling provisions
  • Selectable decimal point (most models)
  • Auto zeroing circuits
  • NEMA 4X/IP65 sealed front bezel
  • Optional custom units overlay with backlight