Red Lion XCDN0000 DeviceNet Option Card
Representative image only.
Red Lion Mfr # XCDN0000 – Item # EW-68516-16
A nexus for industrial data collection and management
  • Converts numerous protocols simultaneously
  • Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
  • Supports up to nine protocols simultaneously (with expansion card)
  • DIN rail mounting
  • Operating termperature 32 to 122°F (0 to 50°C)
PRODUCT OPTIONS
$437.00USD / Each
For better pricing please sign in.
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithData Station Plus operator interfaces
  • DescriptionDeviceNet Option Card
Includes
Hardware packet for mounting unit into panel.
Key Features
  • Converts numerous protocols simultaneously
  • Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
  • Supports up to nine protocols simultaneously (with expansion card)
  • DIN rail mounting
  • Operating termperature 32 to 122°F (0 to 50°C)
More About this Item
The Data Station Plus is designed to act as a nexus for industrial data collection and management. With three built-in serial ports and a 10 Base-T/100 Base-TX Ethernet port, the unit performs protocol conversion, allowing disparate devices to communicate seamlessly with one another. The Data Station Plus functions both as a data management platform and as a PFM host.
The Ethernet port supports up to four protocols simultaneously so even Ethernet-to-Ethernet protocol conversion may be performed. The CompactFlash card allows new configuration files to be loaded into the Data Station Plus or use to data log critical information. The USB port may be used for fast file downloads, or to mount the Data Station’s CompactFlash card as an external drive to your PC.