Reichert 13951000 IR Communications Package for Full-Range Digital Refractometer
Representative image only.
Reichert Mfr # 13951000 – Item # EW-81030-50
Optimize performance of your Reichert refractometers
  • Suitable for use with Reichert Full-Range Digital Refractometers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIR Communications Package for Full-Range Digital Refractometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for use with Reichert Full-Range Digital Refractometers
  • 1 year warranty
More About this Item
Use the refractometer's IR communication port with the IR transceiver and link-up software to upload up to three custom scales. The IR transceiver plugs into the RS-232 port on your PC to allow two-way communication between the refractometer and your PC. Dowload the readings, current calibration data, or up to 225 readings that include time/date stamp, reading value, units, channel name, temperature, and sample number.