Replacement Blade for Benchtop Tubing Cutter 06438-10
Representative image only.
Item # EW-06438-11
Save time and money cutting tubing in house
  • Constructed of durable stainless steel
  • Replacement blade for benchtop tubing cutter 06438-10
PRODUCT OPTIONS
$14.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Minimum ID (in)0.351
  • Minimum ID (mm)8.9
  • DescriptionReplacement Blade for Benchtop Tubing Cutter 06438-10
Key Features
  • Constructed of durable stainless steel
  • Replacement blade for benchtop tubing cutter 06438-10
More About this Item
Steel blade makes right-angle cuts in plastic tubing with an outer diameter of 1/16" to 5/8".