ResinTech VP-17-4012 VP Series Low TOC / Mixed Bed
Representative image only.
ResinTech Mfr # VP-17-4012 – Item # EW-99284-78
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Ultra low TOC activated carbon Standard connection
PRODUCT OPTIONS
$182.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MediaUltra-Low TOC, Mixed-Bed
  • Capacity1360 grains
  • Capacity (grains)1360
  • ConnectionStandard 1" Top x 2" Bottom
  • Compatible withARIES, Unix, Barnstead, Vaponics
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.38
  • Length (in)17
  • DescriptionFilter Cartridge, Ultra Low TOC Mixed-Bed Resin, Standard Connection
  • Warranty1 year
1 Year Warranty
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Ultra low TOC activated carbon Standard connection
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.