ResinTech VPK-4801 VPK Series Cartridge Kit, Pretreatment, High Capacity, (2) High Purity - Equaivalent to Barnstead D4801
Representative image only.
ResinTech Mfr # VPK-4801 – Item # EW-99284-96
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Includes: 1 x VP-17-4205, 1 x VP-17-4203, 2 x VP-17-4010 Equivalent to: D4801
  • 1 year warranty
PRODUCT OPTIONS
$659.12USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MediaActivated Carbon/High Purity, High Capacity, High Purity
  • Capacity490 grains / 2000 gal. / 1590 grains / (2) 1360 grains
  • Capacity (Gal)2000
  • Capacity (grains)1590
  • ConnectionStandard 1\" Top x 2\" Bottom
  • Compatible withARIES, Unix, Barnstead (D4801), Vaponics
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.38
  • Length (in)17
  • DescriptionFilter Cartridge Kit, Activated Carbon/High Purity, High Capacity, Two High Purity, Standard Connection; 4/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
  • Includes: 1 x VP-17-4205, 1 x VP-17-4203, 2 x VP-17-4010 Equivalent to: D4801
  • 1 year warranty
More About this Item
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.