This item is now discontinued. Please browse similar items
RheoSense HA01-01 Viscosity Chip, 0 to 100 cP Flow channel depth: 50 microns
Representative image only.
RheoSense Mfr # HA01-01 – Item # EW-98946-42
Only requires 400 microliter sample size!
SPECS & DESCRIPTION
Specifications & Description
  • Min Viscosity Range (cP)0
  • Max Viscosity Range (cP)100
  • DescriptionViscosity Chip, 0 to 100 cP Flow channel depth: 50 microns
More About this Item
The microfluidic sensors measure viscosity by the drop in pressure that occurs while the sample flows through a small rectangular flow channel.