Ricca Chemical Company 1900-16 Ceric Sulfate, 0.100 Normal (N/10) in 2 Normal Sulfuric Acid (500 mL)

Representative image only.
Ricca Chemical Company Mfr # 1900-16 – Item # EW-86970-87
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
  • CERIC SULFATE, 0.100 Normal

PRODUCT OPTIONS

$67.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.1
  • Purity GradeComponents all ACS or Reagent
  • ClassTitrants
  • ChemistryAcids;Inorganics
  • FormLiquid
  • Volume (mL)500
  • Packaging500 mL Poly Bottle
  • DOT InformationCorrosive Liquid, Acidic, Inorganic, n.o.s., (Sulphuric Acid); Class 8, Packing Group III
  • CAS NumberIncludes 13590-82-4, 7664-93-9 and 7732-18-5
  • UN NumberUN3264
  • Shelf Life (months)24
  • DescriptionCeric Sulfate, 0.100 Normal (N/10) in 2 Normal Sulfuric Acid (500 mL)

Key Features

  • Reagents and solutions to meet your lab and research needs
  • CERIC SULFATE, 0.100 Normal

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat