Ricca Chemical Company 2237-20 Conductivity/TDS Standard, 100 uS/cm (100 umho/cm) at 25C, 20 mL

Representative image only.
Ricca Chemical Company Mfr # 2237-20 – Item # EW-86977-39
  • CONDUCTIVITY STD, 100 uMHOS
  • 20 x 20 mL Pouches
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)20
  • PackagingPouch
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7647-14-5 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionConductivity/TDS Standard, 100 uS/cm (100 umho/cm) at 25C, 20 mL

Key Features

  • CONDUCTIVITY STD, 100 uMHOS
  • 20 x 20 mL Pouches

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat