Ricca Chemical Company 2244.39-1 Conductivity/TDS Standard, 3.9 mS/cm (3900 umho/cm) at 25C, 2012 ppm as NaCl, 4L

Representative image only.
Ricca Chemical Company Mfr # 2244.39-1 – Item # EW-86985-48
Ideal for laboratory and field analysis
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • CONDUCTIVITY STD, 3,900 uM
  • (NaCl in Water)
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)3785
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7647-14-5 and 7732-18-5
  • Shelf Life (months)12
  • DescriptionConductivity/TDS Standard, 3.9 mS/cm (3900 umho/cm) at 25C, 2012 ppm as NaCl, (4 L)

Key Features

  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • CONDUCTIVITY STD, 3,900 uM
  • (NaCl in Water)

More About this Item

Enhance the accuracy of your conductivity meter by calibrating with Ricca conductivity/TDS standards. Choose a conductivity that has a similar value to the solution being measured.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat