Ricca Chemical Company 2249-16 Conductivity/TDS Standard, 100 mS/cm at 25C, 65,750 ppm as NaCl, 500 mL

Representative image only.
Ricca Chemical Company Mfr # 2249-16 – Item # EW-86971-53
  • CONDUCTIVITY STD, 100,000 uM
  • (NaCl in Water)
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeComponents all ACS
  • ClassStandards
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS NumberIncludes 7647-14-5 and 7732-18-5
  • Shelf Life (months)18
  • DescriptionConductivity/TDS Standard, 100 mS/cm at 25C, 65,750 ppm as NaCl, 500 mL

Key Features

  • CONDUCTIVITY STD, 100,000 uM
  • (NaCl in Water)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat